Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAT2, p Tyr689 Antibody (1021D), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB363857 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB363857 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB363857 Supplier Novus Biologicals Supplier No. FAB2890Z

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Monoclonal Antibody

STAT2 Monoclonal antibody specifically detects STAT2 in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry

Specifications

Antigen STAT2
Applications Western Blot, Simple Western, Flow Cytometry
Classification Monoclonal
Clone 1021D
Conjugate DyLight 755
Dilution Western Blot, Simple Western, Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias interferon alpha induced transcriptional activator, ISGF-3, MGC59816, p113, signal transducer and activator of transcription 2, signal transducer and activator of transcription 2, 113kD, signal transducer and activator of transcription 2, 113kDa, STAT113
Host Species Rabbit
Immunogen Phosphopeptide containing the human STAT2 Y690 site
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Phospho Specific, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 6773
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.