Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAT3 Antibody (OTI21E7), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. N274378A700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N274378A700 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N274378A700 Supplier Novus Biologicals Supplier No. NBP274378AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

STAT3 Monoclonal specifically detects STAT3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Knockout Validated.

Specifications

Antigen STAT3
Applications Western Blot, Immunohistochemistry, Immunoassay
Classification Monoclonal
Clone OTI21E7
Conjugate Alexa Fluor 700
Dilution Western Blot, Immunohistochemistry, Knockout Validated
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Acute-phase response factor, APRFMGC16063, DNA-binding protein APRF, FLJ20882, HIES, signal transducer and activator of transcription 3, signal transducer and activator of transcription 3 (acute-phase responsefactor)
Gene Symbols STAT3
Host Species Mouse
Immunogen Recombinant protein expressed in E.coli corresponding to amino acids 154-175 of human STAT3.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cellular Markers, Myeloid derived Suppressor Cell, Phospho Specific, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 6774.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.