Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAT3, p Ser727 Antibody (788335), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB303363 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB303363 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB303363 Supplier Novus Biologicals Supplier No. FAB4934V

Mouse Monoclonal Antibody

STAT3 Monoclonal antibody specifically detects STAT3 in Human samples. It is validated for Western Blot, Flow Cytometry

Specifications

Antigen STAT3
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone 788335
Conjugate Alexa Fluor 405
Dilution Western Blot, Intracellular Staining by Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias Acute-phase response factor, APRFMGC16063, DNA-binding protein APRF, FLJ20882, HIES, signal transducer and activator of transcription 3, signal transducer and activator of transcription 3 (acute-phase responsefactor)
Host Species Mouse
Immunogen Phosphopeptide containing the human STAT3 S727 site
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Myeloid derived Suppressor Cell, Phospho Specific, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 6774
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.