Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STAT3 Rabbit anti-Human, Mouse, PE, Clone: B12, Abnova™
Rabbit Monoclonal Antibody.
$449.50 - $688.00
Specifications
Antigen | STAT3 |
---|---|
Clone | B12 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PE |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-335-964
|
Abnova Corporation
MAB19130 |
10 Reactions |
Each for $449.50
|
|
|||||
89-335-963
|
Abnova Corporation
MAB19129 |
100 Reactions |
Each for $688.00
|
|
|||||
Description
STAT3 Monoclonal antibody specifically detects STAT3 in Human, Mouse samples. It is validated for Flow Cyt.Specifications
STAT3 | |
Flow Cytometry | |
PE | |
Rabbit | |
APRF, FLJ20882, HIES, MGC16063 | |
A synthetic phosphopeptide corresponding to residues surrounding Y705 of human STAT3 | |
Primary | |
signal transducer and activator of transcription 3 (acute-phase response factor) |
B12 | |
Monoclonal | |
Liquid | |
Human, Mouse | |
6774 | |
IgG1 κ | |
Store at 4°C | |
STAT3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title