Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STAT3 Rabbit anti-Human, Mouse, SureLight 488, Clone: B12, Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation MAB19131
Description
STAT3 Monoclonal antibody specifically detects STAT3 in Human, Mouse samples. It is validated for Flow Cyt.Specifications
| STAT3 | |
| Monoclonal | |
| SureLight 488 | |
| STAT3 | |
| Rabbit | |
| 100 Reactions | |
| 6774 | |
| Store at 4°C | |
| IgG1 κ |
| Flow Cytometry | |
| B12 | |
| signal transducer and activator of transcription 3 (acute-phase response factor) | |
| APRF, FLJ20882, HIES, MGC16063 | |
| A synthetic phosphopeptide corresponding to residues surrounding Y705 of human STAT3 | |
| Primary | |
| Human, Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction