Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STAT3 Rabbit anti-Human, Mouse, SureLight 488, Clone: B12, Abnova™
Rabbit Monoclonal Antibody.
$543.54 - $831.18
Specifications
| Antigen | STAT3 |
|---|---|
| Clone | B12 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | SureLight 488 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
89-335-966
|
Abnova Corporation
MAB19132 |
10 Reactions |
Each for $543.54
|
|
|||||
|
89-335-965
|
Abnova Corporation
MAB19131 |
100 Reactions |
Each for $831.18
|
|
|||||
Description
STAT3 Monoclonal antibody specifically detects STAT3 in Human, Mouse samples. It is validated for Flow Cyt.Specifications
| STAT3 | |
| Flow Cytometry | |
| SureLight 488 | |
| Rabbit | |
| APRF, FLJ20882, HIES, MGC16063 | |
| A synthetic phosphopeptide corresponding to residues surrounding Y705 of human STAT3 | |
| Primary | |
| signal transducer and activator of transcription 3 (acute-phase response factor) |
| B12 | |
| Monoclonal | |
| Liquid | |
| Human, Mouse | |
| 6774 | |
| IgG1 κ | |
| Store at 4°C | |
| STAT3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title