Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAT4 Antibody (OTI2F3), PerCP, Novus Biologicals™
SDP

Catalog No. NB274389PCP Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB274389PCP 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB274389PCP Supplier Novus Biologicals Supplier No. NBP274389PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

STAT4 Monoclonal specifically detects STAT4 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen STAT4
Applications Flow Cytometry
Classification Monoclonal
Clone OTI2F3
Conjugate PerCP
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias signal transducer and activator of transcription 4, SLEB11
Gene Symbols STAT4
Host Species Mouse
Immunogen Full length recombinant protein of human STAT4 (NP_003142) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, Phospho Specific, Regulatory Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6775.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.