Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STAT4 Rabbit anti-Human, Mouse, APC, Clone: F6, Abnova™
Rabbit Monoclonal Antibody.
$449.50
Specifications
Antigen | STAT4 |
---|---|
Clone | F6 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | APC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-335-968
|
Abnova Corporation
MAB19134 |
10 Reactions |
Each of 1 for $449.50
|
|
|||||
Description
STAT4 Monoclonal antibody specifically detects STAT4 in Human, Mouse samples. It is validated for Flow Cyt.Specifications
STAT4 | |
Flow Cytometry | |
APC | |
Rabbit | |
SLEB11 | |
A synthetic phosphopeptide corresponding to residues surrounding Y693 of human STAT4 | |
Primary | |
signal transducer and activator of transcription 4 |
F6 | |
Monoclonal | |
Liquid | |
Human, Mouse | |
6775 | |
IgG1 κ | |
Store at 4°C | |
STAT4 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title