Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAT5a/b, p Tyr694, p Tyr699 Antibody (1247C), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB347960 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB347960 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB347960 Supplier Novus Biologicals Supplier No. FAB41901MFV610
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

STAT5a/b Monoclonal antibody specifically detects STAT5a/b in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF

Specifications

Antigen STAT5a/b
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 1247C
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias MGF, STAT5, STAT5A signal transducer and activator of transcription 5A
Host Species Rabbit
Immunogen Phosphopeptide containing human STAT5b Y699 site (amino acid sequence of this peptide is identical to a corresponding region of human STAT5a containing Y694), Accession # P51692
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Phospho Specific, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 6776
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.