Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STAT5A, p Tyr694 Antibody (3F1), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32638750UL
Description
STAT5A Monoclonal antibody specifically detects STAT5A in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
STAT5A | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunohistochemistry | |
3F1 | |
Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 | |
MGF, signal transducer and activator of transcription 5A, STAT5 | |
A synthesized peptide derived from Human STAT5A [p Tyr694] [UniProt P42229] | |
50 μL | |
Cancer, Phospho Specific, Signal Transduction, Transcription Factors and Regulators | |
6776 | |
Store at -20 to -70C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction