Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STAT5A, p Tyr694 Antibody (3F1), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | STAT5A |
---|---|
Clone | 3F1 |
Dilution | Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 |
Applications | Western Blot, ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
STAT5A Monoclonal antibody specifically detects STAT5A in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
STAT5A | |
Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
MGF, signal transducer and activator of transcription 5A, STAT5 | |
A synthesized peptide derived from Human STAT5A [p Tyr694] [UniProt P42229] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
3F1 | |
Western Blot, ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Cancer, Phospho Specific, Signal Transduction, Transcription Factors and Regulators | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
6776 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title