Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAT5A, p Tyr694 Antibody, Novus Biologicals™
SDP

Catalog No. NB10082219 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.05 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10082219 0.05 mL
NB017883 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10082219 Supplier Novus Biologicals Supplier No. NB10082219
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

STAT5a (p Tyr694) Polyclonal specifically detects STAT5a (p Tyr694) in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen STAT5a (p Tyr694)
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100
Formulation PBS (without Mg2+ and Ca2+), pH 7.4, 150 mM NaCl, 50% glycerol with 0.02% Sodium Azide
Gene Accession No. P42229
Gene Alias MGF, signal transducer and activator of transcription 5A, STAT5
Gene Symbols STAT5A
Host Species Rabbit
Immunogen The antiserum was produced against synthesized phosphopeptide derived from human STAT5A around the phosphorylation site of tyrosine 694 (D-G-YP-V-K).
Molecular Weight of Antigen 90 kDa
Purification Method Affinity Purified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6776
Test Specificity Detects endogenous levels of STAT5A only when phosphorylated at tyrosine 694.
Target Species Human, Mouse, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.