Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAT5b Antibody (389215) [Janelia Fluor™ 549], Novus Biologicals™
SDP

Catalog No. NB399399 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB399399 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB399399 Supplier Novus Biologicals Supplier No. FAB1584I
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

STAT5b Monoclonal specifically detects STAT5b in Human samples. It is validated for CyTOF, Flow Cytometry (Intracellular Staining), Gene Knock-Out, Immunoprecipitation, Western Blot.

Specifications

Antigen STAT5b
Applications CyTOF, Flow Cytometry (Intracellular Staining), Gene Knock-Out, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 389215
Conjugate Janelia Fluor 549
Dilution Western Blot, Simple Western, Immunoprecipitation, Intracellular Staining by Flow Cytometry, CyTOF-ready, Knockout Validated
Formulation 50mM Sodium Borate
Gene Alias signal transducer and activator of transcription 5B, STAT5, transcription factor STAT5B
Host Species Mouse
Immunogen Human STAT5b synthetic peptide, MDSQWIPHAQS, Accession # NP_036580
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6777
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.