Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAT5b Antibody (STAT5B/2611), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB012306 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB012306 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB012306 Supplier Novus Biologicals Supplier No. NBP279907DL594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

STAT5b Monoclonal specifically detects STAT5b in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen STAT5b
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone STAT5B/2611
Conjugate DyLight 594
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias signal transducer and activator of transcription 5B, STAT5, transcription factor STAT5B
Gene Symbols STAT5B
Host Species Mouse
Immunogen Recombinant full-length human STAT5b protein (Uniprot: P51692)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific, Signal Transduction
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.