Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAT5b Antibody (STAT5B/2657), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB379424 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB379424 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB379424 Supplier Novus Biologicals Supplier No. NBP308628AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

STAT5b Monoclonal antibody specifically detects STAT5b in Human samples. It is validated for ELISA, Protein Array

Specifications

Antigen STAT5b
Applications ELISA, Protein Array
Classification Monoclonal
Clone STAT5B/2657
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias signal transducer and activator of transcription 5B, STAT5, transcription factor STAT5B
Host Species Mouse
Immunogen Recombinant full-length human STAT5b protein (Uniprot: P51692)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6777
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.