Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAT6 Antibody, Novus Biologicals™
SDP

Catalog No. NB600229 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600229 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600229 Supplier Novus Biologicals Supplier No. NB600229
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

STAT6 Polyclonal specifically detects STAT6 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen STAT6
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:10000, Immunohistochemistry 1:500 - 1:2000, Immunoprecipitation 2-10 μg/mL, Immunohistochemistry-Paraffin 1:500 - 1:2000
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. P42226
Gene Alias D12S1644, EC 2.4.1.227, EC 2.7.7.6, IL-4 Stat, IL-4-STAT, signal transducer and activator of transcription 6, signal transducer and activator of transcription 6, interleukin-4 induced, STAT, interleukin-4 induced, STAT, interleukin4-induced, STAT6B, STAT6C, transcription factor IL-4 STAT
Gene Symbols STAT6
Host Species Rabbit
Immunogen The immunogen recognized by the STAT6 Antibody maps to a region between residue 600 and 650 of human Signal Transducer and Activator of Transcription 6 (NP_003144.3).
Molecular Weight of Antigen 94 kDa
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Immune System Diseases, Immunology, Inflammation, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6778
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, Tasmanian devil, White-tufted-ear marmoset, Crab-eating macaque and Northern white-cheeked gibbon.
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.