Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAT6, p Tyr641 Antibody (1248D), PE, Novus Biologicals™
SDP

Catalog No. NB303836 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB303836 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB303836 Supplier Novus Biologicals Supplier No. FAB3717P

Rabbit Monoclonal Antibody

STAT6 Monoclonal antibody specifically detects STAT6 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry

Specifications

Antigen STAT6
Applications Western Blot, Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 1248D
Conjugate PE
Dilution Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry
Formulation PBS
Gene Alias D12S1644, EC 2.4.1.227, EC 2.7.7.6, IL-4 Stat, IL-4-STAT, signal transducer and activator of transcription 6, signal transducer and activator of transcription 6, interleukin-4 induced, STAT, interleukin-4 induced, STAT, interleukin4-induced, STAT6B, STAT6C, transcription factor IL-4 STAT
Host Species Rabbit
Immunogen Phosphopeptide containing human STAT6 Y641 site., Accession # P42226
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Asthma, Cancer, Cell Biology, Epitope Tags, Immune System Diseases, Immunology, Inflammation, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6778
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.