Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stathmin 1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP176798SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP176798SS 0.025 mL
NBP176798 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP176798SS Supplier Novus Biologicals Supplier No. NBP176798SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Stathmin 1 Polyclonal specifically detects Stathmin 1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated.

Specifications

Antigen Stathmin 1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-1 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry 1:100 - 1:250, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:100 - 1:250
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. P16949
Gene Alias C1orf215, chromosome 1 open reading frame 215, Lag, Leukemia-associated phosphoprotein p18, Metablastin, MGC138869, Oncoprotein 18, Op18, OP18MGC138870, phosphoprotein 19, Phosphoprotein p19, pp17, PP19, PR22, prosolin, Protein Pr22, SMNLAP18FLJ32206, stathmin, stathmin 1, stathmin 1/oncoprotein 18, transmembrane protein C1orf215
Gene Symbols STMN1
Host Species Rabbit
Immunogen A synthetic peptide made to an C-terminal portion of the human Stathmin 1 protein (between residues 100-149) [UniProt P16949]
Molecular Weight of Antigen 17 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Immunology, Microglia Markers, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3925
Test Specificity Human, mouse and rat. Predicted to react in bovine, primate, pig, Chinese Hamster and canine based on 100% sequence identity.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.