Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stathmin-2/STMN2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP149461 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP149461 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP149461 Supplier Novus Biologicals Supplier No. NBP149461
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 34 publications

Stathmin-2/STMN2 Polyclonal specifically detects Stathmin-2/STMN2 in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, In vitro assay, In vivo assay, Knockdown Validated.

Specifications

Antigen Stathmin-2/STMN2
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunohistochemistry (Frozen), In vitro Assay, Immunohistochemistry, KnockDown
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:4000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:4000, Immunohistochemistry-Paraffin reported in scientific literature (PMID 31182472), Immunohistochemistry-Frozen 1:1000, In vitro assay reported in scientific literature (PMID 22726832), In vivo assay reported in scientific literature (PMID 22726832), Immunohistochemistry Whole-Mount reported in scientific literature (PMID 35042776), Knockdown Validated
Formulation Whole antisera with 0.05% Sodium Azide
Gene Alias FLJ34868, FLJ50995, neuron-specific growth-associated protein, Protein SCG10, SCG10, SCG10neuronal growth-associated protein (silencer element), SCGN10stathmin-2, stathmin-like 2, superior cervical ganglia, neural specific 10, Superior cervical ganglion-10 protein
Gene Symbols STMN2
Host Species Rabbit
Immunogen C-terminal peptide of mouse STMN2. [Swiss-Prot P55821]
Molecular Weight of Antigen 22 kDa
Purification Method Unpurified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 11075
Target Species Human, Mouse, Rat, Bovine
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.