Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DESCO™ Statshield™ Metal-In Bag
SDP

Catalog No. p-8874005
Encompass
Click to view available options
Dimensions (L x W):
3 x 5 in.
4 x 6 in.

Metal-In ESD Shielding Bag with low charging Interior and exterior layers.

  • Metal-In Film Laminate 0.0030 in. thick (3 mil)
  • Protects ESD sensitive contents from electrostatic fields and electrostatic discharges (ESD)
  • < 20 nJ Energy Limit Test per ANSI/ESD STM11.31
  • Meets ANSI/ESD S20.20 and ANSI/ESD S541 requirements for ESD shielding packaging inside and outside an ESD Protected Area (EPA)
  • Dissipative Inner and Outer Surfaces Rtt 1 x 104 to < 1 x 1011 ohms per ANSI/ESD STM11.11
  • Allows bag to be used throughout an ESD Protected Area and grounded when placed on a grounded surface or handled by a grounded operator
  • > 10lb Puncture Strength Average per FTMS 101C/4046
  • Provides physical protection for contents
  • Printed with ESD Protective Symbol and Lot Code Information
  • Meets the requirements of ANSI/ESD S541 for quality control purposes
  • Lead-free RoHS compliant
  • Made in the United States of America

Specifications

Certifications/Compliance Meets ANSI/ESD S541
Quantity 100/Pk.
Thickness (English) 3 mil
Dimensions (L x W) 3 x 5 in.
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.