Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STCH Antibody (2F6), PerCP, Novus Biologicals™
SDP

Catalog No. NB308666 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB308666 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB308666 Supplier Novus Biologicals Supplier No. NBP260243PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

STCH Monoclonal antibody specifically detects STCH in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen STCH
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 2F6
Conjugate PerCP
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias heat shock 70 kDa protein 13, heat shock protein 70kDa family, member 13, microsomal stress 70 protein ATPase core, Microsomal stress-70 protein ATPase core, STCHStress-70 protein chaperone microsome-associated 60 kDa protein, stress 70 protein chaperone, microsome-associated, 60kD, stress 70 protein chaperone, microsome-associated, 60kDa
Host Species Mouse
Immunogen Recombinant human STCH (23-471aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6782
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.