Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STCH Antibody, Novus Biologicals™
SDP

Catalog No. NBP220509 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP220509 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP220509 Supplier Novus Biologicals Supplier No. NBP220509
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

STCH Polyclonal specifically detects STCH in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen STCH
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1:100-1:1000
Formulation PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. P48723
Gene Alias heat shock 70 kDa protein 13, heat shock protein 70kDa family, member 13, microsomal stress 70 protein ATPase core, Microsomal stress-70 protein ATPase core, STCHStress-70 protein chaperone microsome-associated 60 kDa protein, stress 70 protein chaperone, microsome-associated, 60kD, stress 70 protein chaperone, microsome-associated, 60kDa
Gene Symbols HSPA13
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human STCH. The exact sequence is proprietary.
Molecular Weight of Antigen 52 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6782
Test Specificity Human. Species reactivity based on sequence identity: Bovine (85%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.