Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stearyldiethanolamine 98.0+%, TCI America™
SDP

Catalog No. O001725G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 10213-78-2
Molecular Formula C22H47NO2
Molecular Weight (g/mol) 357.623
MDL Number MFCD00059003
InChI Key NDLNTMNRNCENRZ-UHFFFAOYSA-N
PubChem CID 61494
IUPAC Name 2-[2-hydroxyethyl(octadecyl)amino]ethanol
SMILES CCCCCCCCCCCCCCCCCCN(CCO)CCO

Specifications

Melting Point 51°C
Color White
Quantity 25 g
Synonym 2,2′-(Octadecylimino)diethanol
Formula Weight 357.62
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Stearyldiethanolamine
EINECSNumber (2)-0353&(7)-0060
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.