Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Stella/Dppa3 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP241089
Description
Stella/Dppa3 Polyclonal specifically detects Stella/Dppa3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.Specifications
Stella/Dppa3 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
DPPA3 | |
Antibody was raised against a 13 amino acid synthetic peptide from near the carboxy terminus of human Stella. The immunogen is located within the last 50 amino acids of Stella. Amino Acid Squence: NGWDPSENARIGN | |
0.1 mg | |
Developmental Biology | |
359787 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
1 mg/mL | |
Western Blot 1-2 μg/mL, ELISA | |
developmental pluripotency associated 3, DPPA3, DPPA3 developmental pluripotency associated 3 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction