Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stella/Dppa3 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB344186 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB344186 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB344186 Supplier Novus Biologicals Supplier No. NBP241089MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Stella/Dppa3 Polyclonal antibody specifically detects Stella/Dppa3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA

Specifications

Antigen Stella/Dppa3
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias developmental pluripotency associated 3, DPPA3, DPPA3 developmental pluripotency associated 3
Host Species Rabbit
Immunogen Antibody was raised against a 13 amino acid synthetic peptide from near the carboxy terminus of human Stella. The immunogen is located within the last 50 amino acids of Stella. Amino Acid Squence: NGWDPSENARIGN
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Developmental Biology
Primary or Secondary Primary
Gene ID (Entrez) 359787
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.