Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stella/Dppa3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP227292SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227292SS 0.025 mg
NBP227292 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227292SS Supplier Novus Biologicals Supplier No. NBP227292SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Stella/Dppa3 Polyclonal specifically detects Stella/Dppa3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Stella/Dppa3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 5ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. Q6W0C5
Gene Alias developmental pluripotency associated 3, DPPA3, DPPA3 developmental pluripotency associated 3
Gene Symbols DPPA3
Host Species Rabbit
Immunogen A portion of amino acids 100-159 of human STELLAR (isoform CRA_b) was used as the immunogen.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Developmental Biology
Primary or Secondary Primary
Gene ID (Entrez) 359787
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_b) and 94% homologous in chimp.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.