Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stella/Dppa3 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$181.90 - $402.00

Specifications

Antigen Stella/Dppa3
Concentration 0.5 mg/mL
Dilution Immunohistochemistry-Paraffin 5ug/ml
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP227292SS
SDP
View Documents
Novus Biologicals
NBP227292SS
0.025 mg
Each for $181.90
Only null left
Add to Cart
 
NBP227292
SDP
View Documents
Novus Biologicals
NBP227292
0.1 mg
Each for $402.00
Only null left
Add to Cart
 
Description

Description

Stella/Dppa3 Polyclonal specifically detects Stella/Dppa3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Stella/Dppa3
Immunohistochemistry-Paraffin 5ug/ml
Polyclonal
Purified
RUO
Human
Q6W0C5
359787
A portion of amino acids 100-159 of human STELLAR (isoform CRA_b) was used as the immunogen.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
0.5 mg/mL
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Developmental Biology
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
developmental pluripotency associated 3, DPPA3, DPPA3 developmental pluripotency associated 3
DPPA3
IgG
Protein A purified
The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_b) and 94% homologous in chimp.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.