Learn More
R&D Systems™ StemXVivo Ectoderm Kit
SDP
Catalog No. SC031B Shop All Novus Products
Change view
Click to view available options
Quantity:
25 Tests
Catalog No. Quantity
SC031B 25 Tests
1 options
This item is not returnable. View return policy
Catalog No. SC031B Supplier R&D Systems™ Supplier No. SC031B
Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
For ectoderm differentiation of human pluripotent stem cells
Embryonic stem (ES) cells are pluripotent stem cells derived from the inner cell mass of pre-implantation embryos. Induced pluripotent stem (iPS) cells can be generated by somatic cell reprogramming following the exogenous expression of specific transcription factors (Oct-3/4, KLF4, SOX2, and c-Myc). These cell types are capable of unlimited, undifferentiated proliferation in vitro and still maintain the capacity to differentiate into a wide variety of somatic cells. In this capacity, pluripotent stem cells have widespread clinical potential for the treatments of heart disease, diabetes, spinal cord injury, and a variety of neurodegenerative disorders.
R&D Systems offers a wide range of products to support pluripotent stem cell culture and differentiation. Mouse embryonic fibroblasts may be used to maintain and expand pluripotent stem cells in an undifferentiated state. We also offer defined culture media, which are specifically optimized for use with human or rodent pluripotent stem cells. In addition, R&D Systems offers a variety of products to assess differentiation status and identify specific stem cell types of interest, including panels of marker antibodies, primer pairs, multi-color flow cytometry kits, and specialized verification kits.
Specifications
Content And Storage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
For Use With (Application) Cell Differentiation
Product Line R&D Systems
Quantity 25 Tests