Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STEP Antibody (23E5), PerCP, Novus Biologicals™
SDP

Catalog No. NB338857 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB338857 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB338857 Supplier Novus Biologicals Supplier No. NB300202PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

STEP Monoclonal antibody specifically detects STEP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)

Specifications

Antigen STEP
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 23E5
Conjugate PerCP
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation PBS
Gene Alias EC 3.1.3.48, FLJ14427, Neural-specific protein-tyrosine phosphatase, protein tyrosine phosphatase, non-receptor type 5 (striatum-enriched), protein-tyrosine phosphatase striatum-enriched, STEPPTPSTEP, Striatum-enriched protein-tyrosine phosphatase, tyrosine-protein phosphatase non-receptor type 5
Host Species Mouse
Immunogen A synthetic peptide of rat STEP46 (amino acids 57-74: MCTPGCNEEGFGYLVSPR)
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Cancer, Cellular Markers, Hypoxia, Neurodegeneration, Neuroscience, Protein Phosphatase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 84867
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.