Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GVS Sterile Cameo Cellulose Acetate Syringe Filters with Glass Prefilters

Catalog No. DGA02025SB Shop All Osmonics Products
Change view
Click to view available options
Pore Size:
0.22 μm
0.45 μm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size
DGA02025SB 0.22 μm
DGA04025S0 0.45 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. DGA02025SB Supplier GVS Supplier No. 1214346

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Ideal for preparing tissue culture media or clarifying biological, protein, enzyme, probe and hybridization buffer solutions

  • 25mm syringe filters with borosilicate glass prefilters and cellulose acetate membranes
  • High throughput, low hold-up volume, negligible protein binding
  • Maximum sample recovery
  • 100% integrity tested acrylic housing
  • Pore size and membrane printed on each filter for easy identification
  • Female locking Luer inlet; male slip Luer outlet
  • Gamma radiation sterilized
  • Packaged in individual blister packs

Specifications

Filter Type Cameo 25GAS
Material Cellulose Acetate
Pore Size 0.22 μm
Sterility Sterile
For Use With (Application) Tissue culture media preparation. Sterile filtration/clarification of: biological fluids; protein, enzyme, probe solutions; hybridization buffers; other aqueous samples.
Hold up Volume (After Air Purge) <60 μL
Inlet Style Female Luer Lok
Outlet Style Male Luer Slip
Pressure Rating 75psi
Quantity 500/Pk.
Temperature Range (English) 131°F
Temperature Range (Metric) 55°C
Tip Material Acrylic
Type Syringe Filter
Diameter (Metric) 25 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.