Learn More
GVS Sterile Cameo Cellulose Acetate Syringe Filters with Glass Prefilters
Shop All Osmonics ProductsDescription
- 25mm syringe filters with borosilicate glass prefilters and cellulose acetate membranes
- High throughput, low hold-up volume, negligible protein binding
- Maximum sample recovery
- 100% integrity tested acrylic housing
- Pore size and membrane printed on each filter for easy identification
- Female locking Luer inlet; male slip Luer outlet
- Gamma radiation sterilized
- Packaged in individual blister packs
Specifications
Specifications
| Filter Type | Syringe Filter |
| Material (Membrane) | Cellulose Acetate |
| Pore Size | 0.45 μm |
| Sterility | Sterile |
| For Use With (Application) | Tissue culture media preparation. Sterile filtration/clarification of: biological fluids, protein, enzyme, probe solutions, hybridization buffers, other aqueous samples. |
| Hold up Volume (After Air Purge) | <60 μL |
| Inlet Style | Female Luer Lock |
| Outlet Style | Male Luer Slip |
| Prefilter | Glass Fiber |
| Pressure Rating | 75 psi |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.