Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stick to Science: Genetic Cards
SCI_ED

Catalog No. s01831b Shop All Si Metric Manufacturing Limited Products
Click to view available options
:
Stick to Science: Genetic Cards
This item is not returnable. View return policy

Catalog No. S01831B

Supplier: Si Metric Manufacturing Limited 50190

This item is not returnable. View return policy

Solve the puzzle of how genes are inherited and sometimes appear or disappear in families.

  • Set of 36 magnetic cards representing males and females with the three possible genetic combinations (affected, carriers or not affected)
  • Investigate all types of inheritance: complete or incomplete dominance, whether autosomal or sex linked
  • Teacher's guide has actual case studies and lesson outlines as well as reproducible student worksheets
  • Grades 7+

Specifications

Grade 7
For Use With (Application) To solve the puzzle of how genes are inherited and sometimes appear or disappear in families
Includes 36 magnetic cards
Product Type Stick to Science: Genetic Card Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.