Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STIM1 Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. N1106047A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1106047A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1106047A70 Supplier Novus Biologicals Supplier No. NB11060547AF750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

STIM1 Polyclonal specifically detects STIM1 in Human, Mouse, Rat, Bovine, Canine samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockdown Validated.

Specifications

Antigen STIM1
Applications Western Blot, Immunocytochemistry, Immunofluorescence, KnockDown
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Western Blot, Immunocytochemistry/Immunofluorescence, Knockdown Validated
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias GOKD11S4896E, stromal interaction molecule 1
Gene Symbols STIM1
Host Species Rabbit
Immunogen STIM1 antibody was made to a synthetic peptide from the C-terminal region (within residues 600-685) of the human STIM1 protein. [UniProt# Q13586]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 6786
Target Species Human, Mouse, Rat, Bovine, Canine
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.