Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STIM1 Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB11060547B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11060547B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11060547B Supplier Novus Biologicals Supplier No. NB11060547B
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

STIM1 Polyclonal specifically detects STIM1 in Human, Mouse, Rat, Bovine, Canine samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockdown Validated.

Specifications

Antigen STIM1
Applications Western Blot, Immunocytochemistry, Immunofluorescence, KnockDown
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot, Immunocytochemistry/Immunofluorescence, Knockdown Validated
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. Q13586
Gene Alias GOKD11S4896E, stromal interaction molecule 1
Gene Symbols STIM1
Host Species Rabbit
Immunogen STIM1 antibody was made to a synthetic peptide from the C-terminal region (within residues 600-685) of the human STIM1 protein. [UniProt# Q13586]
Molecular Weight of Antigen 77 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Plasma Membrane Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6786
Test Specificity Human, mouse, rat, bovine and canine. Immunogen has 93% homology to porcine and 79% homology to Xenopus.
Target Species Human, Mouse, Rat, Bovine, Canine
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.