Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STIM1 Antibody (CDN3H4), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB359839 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB359839 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB359839 Supplier Novus Biologicals Supplier No. NB11055300MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

STIM1 Monoclonal antibody specifically detects STIM1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)

Specifications

Antigen STIM1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CDN3H4
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias GOKD11S4896E, stromal interaction molecule 1
Host Species Mouse
Immunogen Synthetized peptide from C-terminal cytoplasmic part of STIM1. (NM_003156.3).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Plasma Membrane Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6786
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.