Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STIM2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP24417005 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP24417005 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP24417005 Supplier Novus Biologicals Supplier No. NBP2441700.05MG
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

STIM2 Polyclonal specifically detects STIM2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen STIM2
Applications Western Blot, ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 μ/mL, ELISA 1: 10000, Immunohistochemistry-Paraffin
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 30% Glycerol with no preservative
Gene Alias FLJ39527, KIAA1482, stromal interaction molecule 2
Gene Symbols STIM2
Host Species Sheep
Immunogen Stim II affinity purified antibody was prepared from whole sheep serum produced by repeated immunizations with a synthetic peptide near the C-terminus of human STIM2.
Quantity 0.05 mg
Primary or Secondary Primary
Gene ID (Entrez) 57620
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.