Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STING/TMEM173 Antibody (723505) [HRP], Novus Biologicals™
SDP

Catalog No. NB375043 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB375043 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB375043 Supplier Novus Biologicals Supplier No. FAB7169H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

STING/TMEM173 Monoclonal antibody specifically detects STING/TMEM173 in Human samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF

Specifications

Antigen STING/TMEM173
Applications Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 723505
Conjugate HRP
Dilution Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation PBS
Gene Alias endoplasmic reticulum IFN stimulator, Endoplasmic reticulum interferon stimulator, ERIS, FLJ38577, hMITA, hSTING, Mediator of IRF3 activation, MITA, mitochondrial mediator of IRF3 activation, MPYS, NET23, N-terminal methionine-proline-tyrosine-serine plasma membrane tetraspanner, SAVI, Stimulator of interferon genes protein, stimulator of interferon protein, sting, STING-beta, TMEM173, transmembrane protein 173
Host Species Mouse
Immunogen E. coli-derived recombinant human STING/TMEM173, Ala215-Ser379, Accession # Q86WV6
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 340061
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.