Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STING/TMEM173 Antibody, Novus Biologicals™
SDP

Catalog No. NBP238389 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP238389 0.1 mL
NB396850 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP238389 Supplier Novus Biologicals Supplier No. NBP238389
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

STING/TMEM173 Polyclonal specifically detects STING/TMEM173 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen STING/TMEM173
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q86WV6
Gene Alias endoplasmic reticulum IFN stimulator, Endoplasmic reticulum interferon stimulator, ERIS, FLJ38577, hMITA, Mediator of IRF3 activation, MITA, mitochondrial mediator of IRF3 activation, MPYS, NET23, N-terminal methionine-proline-tyrosine-serine plasma membrane tetraspanner, Stimulator of interferon genes protein, sting, STINGhSTING, TMEM173, transmembrane protein 173
Gene Symbols TMEM173
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: YSNSIYELLENGQRAGTCVLEYATPLQTLFAMSQYSQAGFSREDRLEQAKLFCQTLEDILADAPESQNNCR
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 340061
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.