Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STK16 Antibody, DyLight 594, Novus Biologicals™
SDP

Catalog No. NB063664 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB063664 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB063664 Supplier Novus Biologicals Supplier No. NBP299954DL594

Rabbit Polyclonal Antibody

STK16 Polyclonal specifically detects STK16 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen STK16
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 594
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.11.1, FLJ39635, hPSK, MGC16211, MPSK1, MPSKFLJ39635, Myristoylated and palmitoylated serine/threonine-protein kinase, PKL12KRCT, protein kinase expressed in day 12 fetal liver, Protein kinase PKL12, serine/threonine kinase 16, serine/threonine-protein kinase 16, TGF-beta-stimulated factor 1, TSF1, TSF-1
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant STK16 (AAH02618.1; Met1-Ile305).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8576
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.