Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STK31 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110440
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110440 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110440 Supplier Invitrogen™ Supplier No. PA5110440
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GDLSLMSPEL KMGKPASPGS DLYAYGCLLL WLSVQNQEFE INKDGIPKVD QFHLDDKVKS LLCSLICYRS SMTAEQVLNA ECFLMPKEQS VPNPEKDTEY TLYKKEEEIK TENLDKCMEK TRNGEANFDC.

This gene is similar to a mouse gene that encodes a putative protein kinase with a tudor domain, and shows testis-specific expression. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STK31
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene STK31
Gene Accession No. Q99MW1
Gene Alias C330007K24Rik; RGD1564384; serine threonine kinase 31; serine/threonine kinase 31; serine/threonine-protein kinase 31; Serine/threonine-protein kinase NYD-SPK; SGK396; STK31; Sugen kinase 396; TDRD8; testis tissue sperm-binding protein Li 78m; tudor domain containing 8
Gene Symbols STK31
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 890-1019 of human STK31 (NP_113602.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 500120, 77485
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.