Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STK33 Antibody (SR2024), Novus Biologicals™
SDP

Catalog No. NB387918 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB387918 100 μg
NB387917 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB387918 Supplier Novus Biologicals Supplier No. NBP322084100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

STK33 Monoclonal antibody specifically detects STK33 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen STK33
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone SR2024
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:100-1:500
Formulation PBS (pH 7.4), 150 mM NaCl, 50% Glycerol
Gene Alias EC 2.7.11, EC 2.7.11.1, serine/threonine kinase 33, serine/threonine-protein kinase 33
Host Species Rabbit
Immunogen A synthesized peptide derived from human STK33 (Uniprot #: Q9BYT3)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 65975
Target Species Human
Content And Storage Store at -20°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.