Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STK33 Antibody (SR2024), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $413.50
Specifications
Antigen | STK33 |
---|---|
Clone | SR2024 |
Dilution | Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:100-1:500 |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Monoclonal |
Description
STK33 Monoclonal antibody specifically detects STK33 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
STK33 | |
Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:100-1:500 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
EC 2.7.11, EC 2.7.11.1, serine/threonine kinase 33, serine/threonine-protein kinase 33 | |
A synthesized peptide derived from human STK33 (Uniprot #: Q9BYT3) | |
Primary | |
Store at -20°C. Avoid freeze/thaw cycles. |
SR2024 | |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cancer, Protein Kinase | |
PBS (pH 7.4), 150 mM NaCl, 50% Glycerol | |
65975 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title