Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STK33 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501570997
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-0997 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-157-0997 Supplier Bethyl Laboratories, Inc Supplier No. A305676AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Serine Threonine Kinase 33 is normally expressed in embryonic and germ-line cells, which ultimately differentiate into testis, lung, heart, brain and spinal cord. It is critical in both spermatogenesis and organ ontogenesis, and is thus a critical developmental protein. However, in many maligt tissues, STK33 is abnormally expressed, and its appearance in these tissues usually indicates an increase in the cancer cell viability. In cancer cells, STK33 suppresses mitochondrial apoptosis, which precludes normal processes to identify and eliminate cancerous cells.

Specifications

Antigen STK33
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene STK33
Gene Accession No. Q924X7, Q9BYT3
Gene Alias Hypothetical protein FLJ35932
Gene Symbols Stk33
Host Species Rabbit
Immunogen Between 464 and 514
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117229, 65975
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.