Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STK33 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | STK33 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-0997
![]() |
Bethyl Laboratories, Inc
A305676AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt.
Serine Threonine Kinase 33 is normally expressed in embryonic and germ-line cells, which ultimately differentiate into testis, lung, heart, brain and spinal cord. It is critical in both spermatogenesis and organ ontogenesis, and is thus a critical developmental protein. However, in many maligt tissues, STK33 is abnormally expressed, and its appearance in these tissues usually indicates an increase in the cancer cell viability. In cancer cells, STK33 suppresses mitochondrial apoptosis, which precludes normal processes to identify and eliminate cancerous cells.Specifications
STK33 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Hypothetical protein FLJ35932 | |
Between 464 and 514 | |
Primary | |
4° C | |
STK33 |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q924X7, Q9BYT3 | |
117229, 65975 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title