Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STK35 Antibody, Novus Biologicals™
SDP

Catalog No. NB1001393 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001393 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001393 Supplier Novus Biologicals Supplier No. NB1001393
Add to Cart
Add to Cart

Goat Polyclonal Antibody

STK35 Polyclonal specifically detects STK35 in Human, Mouse, Pig samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA, Western Blot.

Specifications

Antigen STK35
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/mL, Flow Cytometry, Immunohistochemistry 10 ug/mL, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin 10 ug/mL, Peptide ELISA Detection limit 1:16000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_543026.
Gene Alias bA550O8.2, CLIK-1, CLIK1Serine/threonine-protein kinase 35 L1, CLP-36 interacting kinase, CLP-36-interacting kinase 1, EC 2.7.11, EC 2.7.11.1, PDIK1, PDLIM1-interacting kinase 1, serine threonine kinase 35 long form, serine/threonine kinase 35, serine/threonine-protein kinase 35, STK35L1
Gene Symbols STK35
Host Species Goat
Immunogen Peptide with sequence ELETRMDQVTCAA corresponding to C-Terminus according to NP_543026.2.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140901
Test Specificity Cross reacts with Human. Expected from sequence similarity: Mouse, Rat, Dog, Pig and Cow.
Target Species Human, Mouse, Pig
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.