Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STK38 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | STK38 |
---|---|
Concentration | 0.71 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6115
![]() ![]() |
Proteintech Group Inc
553351AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody recognizes the C-terminal of STK38.
Stk38,also known as NUCLEAR DBF2-RELATED PROTEIN, contains 1 protein kinase domain that interacts with mob1 and mob2.The homodimeric s100b binds to two molecules of stk38. It is ubiquitously expressed with highest levels observed in peripheral blood leukocytes. Stk38 activated by binding of s100b which releases autoinhibitory n-lobe interactions, enabling atp to bind and the autophosphorylation of ser-281. Thr-444 then undergoes calcium- dependent phosphorylation by an upstream kinase. Interactions between phosphorylated Thr-444 and the n-lobe promote additional structural changes that complete the activation of the kinase. Autoinhibition is also released by the binding of mob1/mobkl1a and mob2/hcca2 to the n-terminal of stk38.Specifications
STK38 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q15208, Q91VJ4 | |
106504, 11329 | |
Peptide | |
Primary | |
-20°C | |
STK38 |
0.71 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
NDR, NDR1, NDR1 protein kinase, Nuclear Dbf2 related kinase 1, serine/threonine kinase 38, STK38 | |
Stk38 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title