Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STK39 Antibody (OTI4E3), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP274406A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP274406A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP274406A94 Supplier Novus Biologicals Supplier No. NBP274406AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

STK39 Monoclonal specifically detects STK39 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen STK39
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI4E3
Conjugate Alexa Fluor 594
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias DCHTsmall intestine SPAK-like kinase, DKFZp686K05124, EC 2.7.11, EC 2.7.11.1, PASK, proline-alanine-rich STE20-related kinase, serine threonine kinase 39, Serine/threonine-protein kinase 39, SPAKserine threonine kinase 39 (STE20/SPS1 homolog, yeast), STE20/SPS1 homolog, STE20/SPS1-related proline-alanine-rich protein kinase, Ste20-like protein kinase, Ste-20-related kinase
Gene Symbols STK39
Host Species Mouse
Immunogen Full length recombinant protein of human STK39 (NP_037365) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 27347.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.