Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STK4 Antibody (025), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB317318 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB317318 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB317318 Supplier Novus Biologicals Supplier No. NBP289912MFV610
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

STK4 Monoclonal antibody specifically detects STK4 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen STK4
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 025
Conjugate mFluor Violet 610 SE
Dilution Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.11, EC 2.7.11.1, kinase responsive to stress 2, KRS2KRS2)), Mammalian STE20-like protein kinase 1, mammalian sterile 20-like 1, MST-1, MST1DKFZp686A2068, serine/threonine kinase 4, serine/threonine-protein kinase 4, Serine/threonine-protein kinase Krs-2, STE20-like kinase MST1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human STK4 (Uniprot#: Q13043-1; Glu2-Phe487).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 6789
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.