Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STK40 Antibody, Novus Biologicals™
SDP

Catalog No. NBP220519 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP220519 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP220519 Supplier Novus Biologicals Supplier No. NBP220519
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

STK40 Polyclonal specifically detects STK40 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen STK40
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000
Formulation PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q8N2I9
Gene Alias 2310004N11Rik, EC 2.7.11, EC 2.7.11.1, MGC4796, Ser/Thr-like kinase, serine/threonine kinase 40, serine/threonine-protein kinase 40, SGK495, SgK495SHIKRP11-268J15.4, SINK-homologous serine/threonine-protein kinase, Sugen kinase 495
Gene Symbols STK40
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human STK40. The exact sequence is proprietary.
Molecular Weight of Antigen 49 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 83931
Test Specificity Human, Mouse. Species reactivity based on sequence identity: Chicken (92%), Rat (96%), Xenopus laevis (88%), Bovine (98%), Xenopus tropicalis (89%).
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.