Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ STOCK2S 26016, >98%
GREENER_CHOICESDP

Catalog No. 557050 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-705-0 50 mg
55-701-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 55-705-0 Supplier Tocris Bioscience™ Supplier No. 5570/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lysine deficient protein kinase (WNK) signaling inhibitor

Lysine deficient protein kinase (WNK) signaling inhibitor (IC50 = 16 μM). Inhibits binding of WNK1 and WNK4 to SPAK and phosphorylation of downstream transporters, NKCC1 and NCC, in mouse vascular smooth muscle and distal convoluted tubule cell cultures exposed to hypotonic shock, respectively.

Chemical Identifiers

CAS 332922-63-1
Molecular Formula C20H19N3O2

Specifications

Quantity 50 mg
Solubility Soluble to 100mM in DMSO and to 50mM in ethan
Formula Weight 333.38
Purity >98%
Molecular Weight (g/mol) 333.391
Chemical Name or Material 7-Ethoxy-N3-(2-furanylmethyl)-3,9-acridinediamine
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.